Assay Details
Target Gene Details
Entrez Gene ID: | 218756 |
Gene Name: | solute carrier family 4, sodium bicarbonate cotransporter, member 7 |
Gene Aliases: |
E430014N10Rik, NBC3, NBCn1, NBCn1-G |
Location: |
Chr.14:14703011-14799944 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc4a7 | NM_001033270.2 | NP_001028442.2 | ||
| AK049322.1 | BAC33682.1 | |||
| AK088400.1 | BAC40330.1 | |||
| GU386352.1 | ||||
| GU386353.2 | ||||
| HM624050.1 | ||||
| HM624051.1 | ||||
| JQ073566.1 | ||||
| JQ344322.1 | ||||
| JQ347261.1 | ||||
| JQ349031.1 | ||||
| JQ349032.1 | ||||
| JQ349033.1 | ||||
| JX254908.1 | ||||
| JX254909.1 | ||||
| JX254910.1 | ||||
| JX254911.1 | ||||
| JX254912.1 | ||||
| JX310364.1 | ||||
| KF279520.1 | ||||
| KF279521.1 |
More Information
Set Membership: |
|

Genomic Map