Assay Details
Target Gene Details
Entrez Gene ID: | 22194 |
Gene Name: | ubiquitin-conjugating enzyme E2E 1 |
Gene Aliases: |
UbcM3, Ubce5, ubcM2 |
Location: |
Chr.14:18282726-18331844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2e1 | NM_009455.3 | NP_033481.1 | ||
| AK090177.1 | BAC41124.1 | |||
| AK148266.1 | BAE28446.1 | |||
| BC003781.1 | AAH03781.1 | |||
| X92665.1 | CAA63353.1 |
More Information
Set Membership: |
|

Genomic Map