Assay Details
Target Gene Details
Entrez Gene ID: | 59011 |
Gene Name: | myozenin 1 |
Gene Aliases: |
2310001N11Rik, AV090278, FATZ, Myoz |
Location: |
Chr.14:20649102-20656540 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myoz1 | NM_021508.3 | 5 | 705 | NP_067483.1 |
| AJ005620.1 | 5 | 645 | CAB76836.1 | |
| AK009077.1 | 5 | 669 | BAB26059.1 | |
| AK041896.1 | 5 | 661 | BAC31092.1 | |
| AY013298.1 | 5 | 623 | AAG38941.1 | |
| BC024660.1 | 5 | 761 | AAH24660.1 |
More Information
Set Membership: |
|

Genomic Map