Assay Details
Target Gene Details
Entrez Gene ID: | 110265 |
Gene Name: | methionine sulfoxide reductase A |
Gene Aliases: |
2310045J23Rik, 6530413P12Rik, MSR-A |
Location: |
Chr.14:64122621-64455903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msra | NM_001253712.1 | NP_001240641.1 | ||
| NM_001253714.1 | NP_001240643.1 | |||
| NM_001253715.1 | NP_001240644.1 | |||
| NM_001253716.1 | NP_001240645.1 | |||
| NM_026322.4 | NP_080598.2 | |||
| AK002356.1 | BAB22035.1 | |||
| AK009822.1 | BAB26522.1 | |||
| AK018338.1 | ||||
| AK030278.1 | ||||
| AK049714.1 | BAC33889.1 | |||
| BB652761.1 | ||||
| BC014738.1 | AAH14738.1 | |||
| BC089311.1 | AAH89311.1 | |||
| BY282106.1 | ||||
| CB953198.1 |
More Information
Set Membership: |
|

Genomic Map