Assay Details
Target Gene Details
Entrez Gene ID: | 57811 |
Gene Name: | retinal G protein coupled receptor |
Gene Aliases: |
- |
Location: |
Chr.14:37034909-37049014 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgr | NM_001301692.1 | NP_001288621.1 | ||
| NM_001301694.1 | NP_001288623.1 | |||
| NM_021340.4 | NP_067315.1 | |||
| AF076930.1 | AAC69836.1 | |||
| AK044450.1 | BAC31926.1 | |||
| AK044649.1 | BAC32017.1 | |||
| AK149272.1 | BAE28782.1 | |||
| BC119055.1 | ||||
| CK622127.1 | ||||
| CK623457.1 | ||||
| CK626179.1 |
More Information
Set Membership: |
|

Genomic Map