Assay Details
Target Gene Details
Entrez Gene ID: | 21933 |
Gene Name: | tumor necrosis factor receptor superfamily, member 10b |
Gene Aliases: |
DR5, KILLER, Ly98, MK, TRAILR2, TRICK2A, TRICK2B, TRICKB |
Location: |
Chr.14:69767472-69784411 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf10b | NM_020275.4 | 5 | 775 | NP_064671.2 |
| AB031081.1 | 5 | 715 | BAA96462.1 | |
| AF176833.1 | 5 | 587 | AAD52656.1 | |
| AK050753.1 | 5 | 776 | BAC34404.1 | |
| AK080069.1 | 5 | 766 | BAC37821.1 | |
| AK145730.1 | 5 | 763 | ||
| AK154993.1 | 5 | 755 | BAE32980.1 | |
| BC065141.1 | 5 | 718 | AAH65141.1 |
More Information
Set Membership: |
|

Genomic Map