Assay Details
Target Gene Details
Entrez Gene ID: | 20387 |
Gene Name: | surfactant associated protein A1 |
Gene Aliases: |
SP-A, Sftp-1, Sftp1 |
Location: |
Chr.14:41131787-41136373 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sftpa1 | NM_023134.4 | NP_075623.2 | ||
| AK004620.1 | BAB23416.1 | |||
| AK004788.1 | BAB23565.1 | |||
| AK011333.1 | BAB27551.1 | |||
| AK075702.1 | ||||
| BC061096.1 | AAH61096.1 |
More Information
Set Membership: |
|

Genomic Map