Assay Details
Target Gene Details
Entrez Gene ID: | 20389 |
Gene Name: | surfactant associated protein C |
Gene Aliases: |
Bricd6, SP-C, SP5, SPC, Sftp-2, Sftp2, pro-SpC |
Location: |
Chr.14:70520941-70524081 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sftpc | NM_011359.2 | NP_035489.2 | ||
| BC061137.1 | AAH61137.1 | |||
| BY773034.1 | ||||
| CJ093163.1 |
More Information
Set Membership: |
|

Genomic Map