Assay Details
Target Gene Details
Entrez Gene ID: | 74843 |
Gene Name: | MSS51 mitochondrial translational activator |
Gene Aliases: |
4833444M15Rik, Zmynd17 |
Location: |
Chr.14:20482862-20498131 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mss51 | NM_029104.1 | 2 | 425 | NP_083380.1 |
| AK014794.1 | 2 | 206 | BAB29556.1 | |
| BC119773.1 | 2 | 318 | ||
| BC119816.1 | 2 | 318 |

Genomic Map