Assay Details
Target Gene Details
Entrez Gene ID: | 219038 |
Gene Name: | tubulin polymerization-promoting protein family member 2 |
Gene Aliases: |
Gm1790, Gm77 |
Location: |
Chr.14:51918425-51920704 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tppp2 | NM_001128634.1 | 2 | 274 | NP_001122106.1 |
| BC049709.1 | 3 | 388 |
More Information
Set Membership: |
|

Genomic Map