Assay Details
Target Gene Details
Entrez Gene ID: | 66597 |
Gene Name: | tripartite motif-containing 13 |
Gene Aliases: |
3110001L12Rik, CAR, LEU5, RNF77, Rfp2 |
Location: |
Chr.14:61598226-61605946 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim13 | NM_001164220.1 | NP_001157692.1 | ||
| NM_023233.3 | NP_075722.1 | |||
| AF220129.1 | AAG53502.1 | |||
| AK013959.2 | BAC25419.1 | |||
| AK017850.1 | BAB30973.1 | |||
| AK162129.1 | BAE36743.1 | |||
| BC138576.1 | ||||
| BC145915.1 | ||||
| BM933374.1 |
More Information
Set Membership: |
|

Genomic Map