Assay Details
Target Gene Details
Entrez Gene ID: | 93834 |
Gene Name: | pellino 2 |
Gene Aliases: |
AW047589, BB129927 |
Location: |
Chr.14:48120778-48308103 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Peli2 | NM_033602.2 | NP_291080.2 | ||
| AF302504.1 | AAG15392.1 | |||
| AK030564.1 | BAC27024.1 | |||
| AK033815.1 | BAC28485.1 | |||
| AK045515.1 | ||||
| BC027062.1 | AAH27062.1 |
More Information
Set Membership: |
|

Genomic Map