Assay Details
Target Gene Details
Entrez Gene ID: | 11429 |
Gene Name: | aconitase 2, mitochondrial |
Gene Aliases: |
Aco-2, Aco3, D10Wsu183e |
Location: |
Chr.15:81872463-81915137 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aco2 | NM_080633.2 | NP_542364.1 | ||
| AK143917.1 | BAE25602.1 | |||
| AK144207.1 | BAE25770.1 | |||
| AK145511.1 | BAE26479.1 | |||
| AK150027.1 | BAE29252.1 | |||
| AK150954.1 | ||||
| AK165411.1 | BAE38169.1 | |||
| AK188036.1 | ||||
| AK213382.1 | ||||
| BC004645.1 | AAH04645.1 | |||
| BC094462.1 | AAH94462.1 | |||
| BI987806.1 | ||||
| BY732916.1 |
More Information
Set Membership: |
|

Genomic Map