Assay Details
Target Gene Details
Entrez Gene ID: | 67712 |
Gene Name: | solute carrier family 25, member 37 |
Gene Aliases: |
1700020E22Rik, 4930513O14Rik, 4930526G11Rik, AI848481, C330015G08Rik, Mfrn, Mfrn1, Mscp, frascati, mitoferrin |
Location: |
Chr.14:69241848-69285103 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a37 | NM_026331.3 | NP_080607.2 | ||
| AF288621.1 | AAL23859.1 | |||
| AF361699.1 | 2 | 2308 | AAL27990.1 | |
| AK006155.1 | ||||
| AK015770.1 | BAB29969.1 | |||
| AK028588.1 | 1 | 1666 | BAC26021.1 | |
| AK046008.1 | ||||
| AK050414.1 | BAC34243.1 | |||
| AK162615.1 | 1 | 461 | BAE36991.1 | |
| BC040399.1 | 1 | 422 | AAH40399.1 |

Genomic Map