Assay Details
Target Gene Details
Entrez Gene ID: | 52535 |
Gene Name: | methyltransferase like 17 |
Gene Aliases: |
2310032K15Rik, D14Ertd209e, Mett11d1 |
Location: |
Chr.14:51884842-51891868 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl17 | NM_001029990.1 | 5 | 602 | NP_001025161.1 |
| AK009576.1 | 5 | 516 | ||
| AK087101.1 | 5 | 534 | ||
| AK155096.1 | 5 | 521 | BAE33043.1 | |
| AK156895.1 | 4 | 652 | ||
| AK172330.1 | 4 | 652 | ||
| BC138944.1 | 5 | 555 | ||
| CF172670.1 | 5 | 602 |
More Information
Set Membership: |
|

Genomic Map