Assay Details
Target Gene Details
Entrez Gene ID: | 52535 |
Gene Name: | methyltransferase like 17 |
Gene Aliases: |
2310032K15Rik, D14Ertd209e, Mett11d1 |
Location: |
Chr.14:51884842-51891868 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl17 | NM_001029990.1 | NP_001025161.1 | ||
| AK009576.1 | ||||
| AK087101.1 | ||||
| AK155096.1 | BAE33043.1 | |||
| AK156895.1 | ||||
| AK172330.1 | ||||
| BC138944.1 |
More Information
Set Membership: |
|

Genomic Map