Assay Details
Target Gene Details
Entrez Gene ID: | 21834 |
Gene Name: | thyroid hormone receptor beta |
Gene Aliases: |
Nr1a2, T3R[b], T3Rbeta, Thrb1, Thrb2, c-erbAbeta |
Location: |
Chr.14:17659218-18040097 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thrb | NM_001113417.1 | NP_001106888.1 | ||
| AK158826.1 | BAE34683.1 |
More Information
Set Membership: |
|

Genomic Map