Assay Details
Target Gene Details
Entrez Gene ID: | 105446 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
1810008P16Rik, 5730544D12Rik, AA959850 |
Location: |
Chr.14:55672235-55687634 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gmpr2 |
Target Gene Details
Entrez Gene ID: | 28113 |
Gene Name: | Terf1 (TRF1)-interacting nuclear factor 2 |
Gene Aliases: |
AW552114, D14Wsu146e, Tin2 |
Location: |
Chr.14:55676208-55682624 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tinf2 | NM_145705.3 | NP_663751.2 | ||
| AF214013.1 | AAF23504.1 | |||
| AF518764.1 | AAN32986.1 | |||
| AK011154.1 | ||||
| AK016456.1 | ||||
| AK028952.1 | ||||
| AK090324.1 | ||||
| AK144592.1 | BAE25954.1 | |||
| AK186139.1 | ||||
| BB630902.1 | ||||
| BC017142.1 | 1 | 442 | ||
| BC030347.1 | AAH30347.1 |
More Information
Set Membership: |
|

Genomic Map