Assay Details
Target Gene Details
Entrez Gene ID: | 68966 |
Gene Name: | neuroguidin, EIF4E binding protein |
Gene Aliases: |
1500001L15Rik, Ngd, neuroguidin |
Location: |
Chr.14:55015454-55024138 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ngdn | NM_026890.2 | 8 | 743 | NP_081166.1 |
| AK005098.1 | 8 | 625 | BAB23816.1 | |
| BC023770.1 | 7 | 628 | AAH23770.1 | |
| BC052790.1 | 8 | 681 | AAH52790.1 |
More Information
Set Membership: |
|

Genomic Map