Assay Details
Target Gene Details
Entrez Gene ID: | 59007 |
Gene Name: | N-glycanase 1 |
Gene Aliases: |
1110002C09Rik, PNGase, Png1 |
Location: |
Chr.14:16249314-16311926 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ngly1 | NM_021504.3 | 7 | 1122 | NP_067479.2 |
| AF250927.1 | 7 | 1109 | AAF74723.1 | |
| AK028248.1 | 7 | 1123 | BAC25839.1 | |
| AK148931.1 | 7 | 1125 | ||
| AK169205.1 | 7 | 1110 | ||
| BC028961.1 | 7 | 1076 | AAH28961.1 |
More Information
Set Membership: |
|

Genomic Map