Assay Details
Target Gene Details
Entrez Gene ID: | 239552 |
Gene Name: | apolipoprotein L 8 |
Gene Aliases: |
9830006J20Rik, APOL-II, Apol2 |
Location: |
Chr.15:77745109-77757044 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apol8 | NM_001081970.1 | NP_001075439.1 | ||
| AK036408.1 | BAC29418.1 | |||
| AK046043.1 | ||||
| BC129972.1 |
More Information
Set Membership: |
|

Genomic Map