Assay Details
Target Gene Details
Entrez Gene ID: | 14573 |
Gene Name: | glial cell line derived neurotrophic factor |
Gene Aliases: |
AI385739 |
Location: |
Chr.15:7810048-7837580 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdnf | NM_001301332.1 | 1 | 1012 | NP_001288261.1 |
| NM_010275.3 | 1 | 1012 | NP_034405.1 | |
| BC119031.1 | ||||
| CJ061678.1 | ||||
| D88264.1 | 1 | 214 | BAA13566.1 | |
| U37459.1 | AAB18672.1 | |||
| U75532.1 | 1 | 129 | AAB18343.1 |
More Information
Set Membership: |
|

Genomic Map