Assay Details
Target Gene Details
Entrez Gene ID: | 13992 |
Gene Name: | KH domain containing, RNA binding, signal transduction associated 3 |
Gene Aliases: |
Etle, SLM-2, Salp, Slm2, T-STAR |
Location: |
Chr.15:68928405-69101163 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Khdrbs3 | NM_010158.2 | NP_034288.2 | ||
| AF079763.1 | AAC31753.1 | |||
| AF099092.1 | AAC72396.1 | |||
| AK014353.1 | ||||
| AK132792.1 | ||||
| AK160564.1 | ||||
| BC031507.1 | AAH31507.1 | |||
| BC057577.1 | AAH57577.1 |
More Information
Set Membership: |
|

Genomic Map