Assay Details
Target Gene Details
Entrez Gene ID: | 14114 |
Gene Name: | fibulin 1 |
Gene Aliases: |
- |
Location: |
Chr.15:85205944-85286294 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbln1 | NM_010180.2 | NP_034310.2 | ||
| AK035388.1 | BAC29054.1 | |||
| AK083573.1 | ||||
| AK085989.1 | ||||
| AK086451.1 | BAC39669.1 | |||
| AK137511.1 | ||||
| AK159646.1 | BAE35258.1 | |||
| AK183901.1 | ||||
| AK188770.1 | ||||
| BC007140.1 | AAH07140.1 | |||
| EU543224.1 | ||||
| X70854.1 | CAA50207.1 |
More Information
Set Membership: |
|

Genomic Map