Assay Details
Target Gene Details
Entrez Gene ID: | 12124 |
Gene Name: | BCL2-interacting killer |
Gene Aliases: |
Biklk, Blk, Nbk |
Location: |
Chr.15:83526792-83544635 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bik | NM_007546.2 | 2 | 184 | NP_031572.1 |
| AF048838.1 | 2 | 183 | AAC40079.1 | |
| AK144937.1 | 2 | 185 | BAE26145.1 | |
| BC003732.1 | 2 | 579 | AAH03732.1 |

Genomic Map