Assay Details
Target Gene Details
Entrez Gene ID: | 66398 |
Gene Name: | COMM domain containing 5 |
Gene Aliases: |
2310065H03Rik, AI854466, D15Ertd81e, Hcarg |
Location: |
Chr.15:76899941-76901297 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Commd5 | NM_025536.2 | NP_079812.1 | ||
| AK010045.1 | BAB26663.1 | |||
| AK013148.1 | 1 | 53 | ||
| BC025891.1 | AAH25891.1 |
More Information
Set Membership: |
|

Genomic Map