Assay Details
Target Gene Details
Entrez Gene ID: | 17117 |
Gene Name: | alpha-methylacyl-CoA racemase |
Gene Aliases: |
Macr1 |
Location: |
Chr.15:10981756-10996624 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amacr | NM_008537.4 | 2 | 376 | NP_032563.2 |
| AK002401.1 | 2 | 344 | BAB22072.2 | |
| AK048249.1 | 2 | 377 | BAC33284.1 | |
| AK160586.1 | 2 | 379 | BAE35893.1 | |
| BC015825.1 | 2 | 304 | AAH15825.1 | |
| U89906.1 | 2 | 284 | AAB72146.1 |
More Information
Set Membership: |
|

Genomic Map