Assay Details
Target Gene Details
Entrez Gene ID: | 67040 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 17 |
Gene Aliases: |
2610007K22Rik, A430025E01Rik, AI047725, C80929, Gm926, p72 |
Location: |
Chr.15:79527696-79546764 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx17 | NM_001040187.1 | 7 | 1218 | NP_001035277.1 |
| NM_199079.1 | 7 | 1037 | NP_951061.1 | |
| NM_199080.2 | 7 | 1218 | NP_951062.1 | |
| AK039888.1 | 7 | 1037 | BAC30474.1 | |
| AK151061.1 | 7 | 993 | BAE30078.1 | |
| AK152749.1 | 7 | 994 | ||
| AK152831.1 | 7 | 993 | BAE31531.1 | |
| AK161001.1 | 7 | 1034 | ||
| AK161012.1 | 7 | 1033 | BAE36146.1 | |
| BC062910.1 | 7 | 1023 | AAH62910.1 | |
| BC096036.1 | 7 | 1218 | AAH96036.1 |

Genomic Map