Assay Details
Target Gene Details
Entrez Gene ID: | 14042 |
Gene Name: | exostoses (multiple) 1 |
Gene Aliases: |
AA409028 |
Location: |
Chr.15:53068261-53346183 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ext1 | NM_010162.2 | 9 | 2627 | NP_034292.2 |
| AK132327.1 | 9 | 2604 | BAE21106.1 | |
| BC004741.1 | 9 | 1991 | AAH04741.1 | |
| U78539.1 | 9 | 2131 | AAB41728.1 | |
| X96639.1 | 9 | 2398 | CAA65443.1 |
More Information
Set Membership: |
|

Genomic Map