Assay Details
Target Gene Details
Entrez Gene ID: | 66120 |
Gene Name: | FK506 binding protein 11 |
Gene Aliases: |
1110002O23Rik, FKBP-11 |
Location: |
Chr.15:98724366-98728198 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp11 | NM_024169.3 | NP_077131.2 | ||
| AK003331.1 | BAB22719.1 | |||
| AK007813.1 | ||||
| AK019132.1 | BAB31559.1 | |||
| BC037596.1 | AAH37596.1 |
More Information
Set Membership: |
|

Genomic Map