Assay Details
Target Gene Details
Entrez Gene ID: | 11732 |
Gene Name: | progressive ankylosis |
Gene Aliases: |
Ankh, D15Ertd221e, ank, mKIAA1581 |
Location: |
Chr.15:27466677-27594909 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ank | NM_020332.4 | 8 | 1287 | NP_065065.3 |
| AF001533.2 | 6 | 633 | AAB65654.2 | |
| AF274752.1 | 8 | 990 | AAF88038.1 | |
| AK083135.1 | 8 | 1288 | BAC38778.1 | |
| AK086014.1 | 1 | 158 | ||
| AK148070.1 | 8 | 1060 | BAE28324.1 | |
| BC054379.1 | 8 | 1194 | AAH54379.1 | |
| DQ832285.1 | 8 | 1287 |

Genomic Map