Assay Details
Target Gene Details
Entrez Gene ID: | 12465 |
Gene Name: | chaperonin containing Tcp1, subunit 5 (epsilon) |
Gene Aliases: |
Ccte, TCPE, mKIAA0098 |
Location: |
Chr.15:31590813-31601804 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cct5 | NM_007637.2 | NP_031663.1 | ||
| AK077753.1 | ||||
| AK088184.1 | BAC40194.1 | |||
| AK129056.1 | BAC97866.1 | |||
| AK133857.1 | BAE21891.1 | |||
| AK136493.1 | BAE23007.1 | |||
| AK145445.1 | BAE26441.1 | |||
| AK146405.1 | BAE27146.1 | |||
| AK146425.1 | BAE27159.1 | |||
| AK150390.1 | BAE29518.1 | |||
| AK153911.1 | BAE32250.1 | |||
| AK167895.1 | BAE39906.1 | |||
| AK217822.1 | ||||
| BC094427.1 | AAH94427.1 | |||
| Z31555.1 | CAA83430.1 |
More Information
Set Membership: |
|

Genomic Map