Assay Details
Target Gene Details
Entrez Gene ID: | 66335 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit C1 |
Gene Aliases: |
1700025B18Rik, U13839 |
Location: |
Chr.15:38661904-38692444 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v1c1 | NM_025494.3 | 2 | 257 | NP_079770.2 |
| AB088407.1 | 2 | 204 | BAC57953.1 | |
| AK006323.1 | 2 | 208 | BAB24526.1 | |
| AK075779.1 | 2 | 229 | BAC35953.1 | |
| AK129007.1 | 2 | 217 | ||
| AK151237.1 | 2 | 218 | BAE30229.1 | |
| AK168914.1 | 2 | 208 | BAE40727.1 | |
| AK197353.1 | 2 | 97 | ||
| BB869068.1 | 2 | 258 | ||
| BC010217.1 | 2 | 205 | AAH10217.1 | |
| U13839.1 | 2 | 199 | AAC83084.1 |
More Information
Set Membership: |
|

Genomic Map