Assay Details
Target Gene Details
Entrez Gene ID: | 55944 |
Gene Name: | eukaryotic translation initiation factor 3, subunit D |
Gene Aliases: |
66/67kDa, AA407891, Eif3s7, eIF3p66 |
Location: |
Chr.15:77958998-77971088 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3d | NM_018749.2 | NP_061219.2 | ||
| AB012580.1 | BAA25327.1 | |||
| AK045228.1 | BAC32270.1 | |||
| AK049767.1 | BAC33910.1 | |||
| AK137428.1 | 2 | 549 | ||
| AK145689.1 | BAE26591.1 | |||
| AK168256.1 | BAE40204.1 | |||
| AK185859.1 | ||||
| AK193840.1 | ||||
| BC089020.1 | AAH89020.1 |
More Information
Set Membership: |
|

Genomic Map