Assay Details
Target Gene Details
Entrez Gene ID: | 170787 |
Gene Name: | histone deacetylase 10 |
Gene Aliases: |
AW548891, Hd10 |
Location: |
Chr.15:89123303-89128700 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hdac10 | NM_199198.2 | 1 | 236 | NP_954668.2 |
| NR_028447.1 | 1 | 236 | ||
| NR_028448.1 | 1 | 236 | ||
| NR_028449.1 | 1 | 236 | ||
| AK040235.1 | 1 | 188 | ||
| AK045028.1 | 1 | 217 | ||
| AK154045.1 | 1 | 200 | BAE32336.1 | |
| BC013700.1 | 1 | 189 | ||
| BC064018.1 | 1 | 163 | AAH64018.1 | |
| BU706947.1 |

Genomic Map