Assay Details
Target Gene Details
Entrez Gene ID: | 97998 |
Gene Name: | DEP domain containing MTOR-interacting protein |
Gene Aliases: |
Depdc6 |
Location: |
Chr.15:55112317-55259273 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Deptor | NM_001037937.3 | NP_001033026.1 | ||
| NM_145470.3 | NP_663445.2 | |||
| AK139888.1 | BAE24172.1 | |||
| AK145208.1 | BAE26298.1 | |||
| AK188097.1 | ||||
| AK220300.1 | BAD90225.1 |
More Information
Set Membership: |
|

Genomic Map