Assay Details
Target Gene Details
Entrez Gene ID: | 16341 |
Gene Name: | eukaryotic translation initiation factor 3, subunit E |
Gene Aliases: |
48kDa, Eif3s6, Int6, eIF3-p46, eIF3-p48 |
Location: |
Chr.15:43250040-43282736 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3e | NM_008388.2 | NP_032414.1 | ||
| AK002576.1 | ||||
| AK011436.1 | BAB27621.1 | |||
| AK083874.1 | 3 | 3163 | ||
| AK088104.1 | ||||
| AK088269.1 | ||||
| AK145059.1 | BAE26211.1 | |||
| AK146936.1 | BAE27546.1 | |||
| AK161650.1 | BAE36512.1 | |||
| AK165929.1 | BAE38465.1 | |||
| AK181127.1 | ||||
| BC029177.1 | AAH29177.1 |
More Information
Set Membership: |
|

Genomic Map