Assay Details
Target Gene Details
Entrez Gene ID: | 68135 |
Gene Name: | eukaryotic translation initiation factor 3, subunit H |
Gene Aliases: |
1110008A16Rik, 40kD, 9430017H16Rik, EIF3-P40, EIF3-gamma, Eif3s3 |
Location: |
Chr.15:51786560-51865515 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3h | NM_080635.1 | NP_542366.1 | ||
| AK003560.1 | BAC25040.1 | |||
| AK020033.1 | ||||
| AK088436.1 | BAC40351.1 | |||
| AK146200.1 | BAE26973.1 | |||
| AK149081.1 | ||||
| AK159113.1 | BAE34829.1 | |||
| AK160335.1 | ||||
| AK167651.1 | BAE39702.1 | |||
| AK167815.1 | BAE39841.1 | |||
| AK167844.1 | BAE39864.1 | |||
| AK167928.1 | BAE39932.1 | |||
| AK168108.1 | BAE40079.1 | |||
| AK168123.1 | ||||
| AK168259.1 | BAE40207.1 | |||
| AK168274.1 | BAE40220.1 | |||
| AK168553.1 | BAE40427.1 | |||
| AK169416.1 | BAE41162.1 | |||
| AK200513.1 | ||||
| BC014755.1 | AAH14755.1 | |||
| BC086915.1 | AAH86915.1 | |||
| BC091728.1 | AAH91728.1 |
More Information
Set Membership: |
|

Genomic Map