Assay Details
Target Gene Details
Entrez Gene ID: | 223691 |
Gene Name: | eukaryotic translation initiation factor 3, subunit L |
Gene Aliases: |
0610011H21Rik, D15N1e, Eif3eip, Eif3ip, Eif3s6ip, HSP-66Y, PAF67 |
Location: |
Chr.15:79075223-79094400 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3l | NM_145139.2 | 13 | 1746 | NP_660121.2 |
| AJ310346.1 | 13 | 1756 | CAC84554.1 | |
| AK032936.1 | 13 | 1734 | BAC28090.1 | |
| AK075584.1 | 13 | 1752 | BAC35837.1 | |
| AK150895.1 | 13 | 1741 | BAE29941.1 | |
| AK151175.1 | 13 | 1741 | BAE30177.1 | |
| AK167640.1 | 13 | 1741 | BAE39691.1 | |
| AK168009.1 | 13 | 1744 | BAE39996.1 | |
| AK168295.1 | 13 | 1743 | BAE40238.1 | |
| BC024463.1 | 13 | 1746 | AAH24463.1 |
More Information
Set Membership: |
|

Genomic Map