Assay Details
Target Gene Details
Entrez Gene ID: | 80287 |
Gene Name: | apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3 |
Gene Aliases: |
Arp3, BC003314, Cem15, Rfv3 |
Location: |
Chr.15:79892408-79908905 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apobec3 | NM_001160415.1 | 1 | 80 | NP_001153887.1 |
| NM_030255.3 | 1 | 80 | NP_084531.2 | |
| AB461446.1 | ||||
| AB461447.1 | ||||
| AB461448.1 | ||||
| AB461449.1 | ||||
| AK044394.1 | 1 | 40 | BAC31901.1 | |
| AK049256.1 | 1 | 41 | ||
| AK153719.1 | 1 | 30 | BAE32154.1 | |
| AK167260.1 | 1 | 39 | ||
| BC003314.1 | 1 | 62 | AAH03314.1 | |
| BY228556.1 | 1 | 80 | ||
| EU707568.1 | ||||
| EU707569.1 | ||||
| EU707570.1 | ||||
| EU707571.1 | ||||
| GQ871501.1 | ||||
| GQ871503.1 |
Target Gene Details
Entrez Gene ID: | 109361 |
Gene Name: | RIKEN cDNA D730005E14 gene |
Gene Aliases: |
- |
Location: |
Chr.15:79889532-79893138 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D730005E14Rik | NR_030675.1 | 1 | 652 | |
| AK035732.1 | 1 | 654 | ||
| AK143849.1 | 1 | 636 | BAE25568.1 |
More Information
Set Membership: |
|

Genomic Map