Assay Details
Target Gene Details
Entrez Gene ID: | 80287 |
Gene Name: | apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3 |
Gene Aliases: |
Arp3, BC003314, Cem15, Rfv3 |
Location: |
Chr.15:79892408-79908905 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apobec3 | NM_001160415.1 | NP_001153887.1 | ||
| NM_030255.3 | NP_084531.2 | |||
| AB461446.1 | ||||
| AB461447.1 | ||||
| AB461448.1 | ||||
| AB461449.1 | ||||
| AK044394.1 | BAC31901.1 | |||
| AK049256.1 | ||||
| AK153719.1 | BAE32154.1 | |||
| AK167260.1 | 2 | 236 | ||
| AK167986.1 | BAE39977.1 | |||
| BC003314.1 | AAH03314.1 | |||
| BY228556.1 | ||||
| EU707568.1 | ||||
| EU707569.1 | ||||
| EU707570.1 | ||||
| EU707571.1 | ||||
| GQ871501.1 | ||||
| GQ871502.1 | ||||
| GQ871503.1 |
More Information
Set Membership: |
|

Genomic Map