Assay Details
Target Gene Details
Entrez Gene ID: | 666661 |
Gene Name: | apolipoprotein L 7c pseudogene |
Gene Aliases: |
- |
Location: |
Chr.15:77618968-77627398 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm8221 | NR_033577.1 | 1 | 302 |

Genomic Map