Assay Details
Target Gene Details
Entrez Gene ID: | 71939 |
Gene Name: | apolipoprotein L 6 |
Gene Aliases: |
2310076O14Rik |
Location: |
Chr.15:77044734-77057108 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apol6 | NM_001163621.1 | NP_001157093.1 | ||
| NM_028010.1 | NP_082286.1 | |||
| AK010208.1 | BAB26769.1 | |||
| AK144563.1 | BAE25940.1 |
Target Gene Details
Entrez Gene ID: | 17189 |
Gene Name: | myoglobin |
Gene Aliases: |
AI325109 |
Location: |
Chr.15:77015487-77050668 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mb | NM_013593.3 | NP_038621.2 | ||
| BC025172.1 | AAH25172.1 |
More Information
Set Membership: |
|

Genomic Map