Assay Details
Target Gene Details
Entrez Gene ID: | 75761 |
Gene Name: | apolipoprotein L 7a |
Gene Aliases: |
9130022K13Rik, Apol3 |
Location: |
Chr.15:77388217-77399110 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apol7a | NM_001164640.1 | NP_001158112.1 | ||
| NM_029419.2 | NP_083695.2 | |||
| AK018646.1 | BAB31325.1 | |||
| AK085956.1 | BAC39578.1 | |||
| BC139137.1 | ||||
| BC139138.1 |

Genomic Map