Assay Details
Target Gene Details
Entrez Gene ID: | 16520 |
Gene Name: | potassium inwardly-rectifying channel, subfamily J, member 4 |
Gene Aliases: |
IRK3, Kcnf2, MB-IRK3 |
Location: |
Chr.15:79483714-79511883 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnj4 | NM_008427.4 | NP_032453.3 | ||
| BC120593.1 | ||||
| BC137594.1 | ||||
| S71382.1 | AAB31087.1 | |||
| U11075.1 | AAA53241.1 |
More Information
Set Membership: |
|

Genomic Map