Assay Details
Target Gene Details
Entrez Gene ID: | 57274 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 8 |
Gene Aliases: |
Mct3 |
Location: |
Chr.15:79251016-79254748 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a8 | NM_020516.2 | NP_065262.1 | ||
| AF019111.2 | AAB70582.2 | |||
| BC018216.1 | AAH18216.1 |
More Information
Set Membership: |
|

Genomic Map