Assay Details
Target Gene Details
Entrez Gene ID: | 170790 |
Gene Name: | megalencephalic leukoencephalopathy with subcortical cysts 1 homolog (human) |
Gene Aliases: |
AW048630, BB074274, Kiaa0027-hp, LVM, MLC, VL, WKL1, mKIAA0027 |
Location: |
Chr.15:88955884-88982687 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mlc1 | NM_133241.2 | NP_573504.1 | ||
| AF449425.1 | AAL66020.1 | |||
| AK030396.1 | BAC26941.1 | |||
| AK043757.1 | BAC31643.1 | |||
| AK122190.1 | BAC65472.1 | |||
| BC024719.1 | AAH24719.1 |

Genomic Map