Assay Details
Target Gene Details
Entrez Gene ID: | 20524 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, peroxisomal membrane protein), member 17 |
Gene Aliases: |
34kDa, 47kDa, PMP34, Pmp47 |
Location: |
Chr.15:81318921-81360765 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a17 | NM_011399.3 | NP_035529.1 | ||
| AJ006341.1 | CAA06984.1 | |||
| AK002388.1 | BAB22062.1 | |||
| AK003672.1 | ||||
| BC008571.1 | AAH08571.1 | |||
| BC011292.1 | AAH11292.1 |

Genomic Map