Assay Details
Target Gene Details
Entrez Gene ID: | 26942 |
Gene Name: | sperm associated antigen 1 |
Gene Aliases: |
tpis |
Location: |
Chr.15:36175483-36235621 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spag1 | NM_012031.3 | NP_036161.2 | ||
| AF181252.1 | AAF06160.1 | |||
| AK032601.1 | BAC27944.1 | |||
| BC046313.1 | AAH46313.1 |

Genomic Map