Assay Details
Target Gene Details
Entrez Gene ID: | 626596 |
Gene Name: | regulator of G-protein signalling 22 |
Gene Aliases: |
- |
Location: |
Chr.15:35960511-36140630 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs22 | NM_001195748.1 | NP_001182677.1 |

Genomic Map